Skip to Content
Merck
All Photos(1)

Key Documents

304700

Ethylenediamine-1,2-14C dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N14CH214CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
133.02
MDL number:

storage temp.

2-8°C

SMILES string

Cl.N[14CH2][14CH2]N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service