Skip to Content
Merck
All Photos(1)

Key Documents

30360

Sigma-Aldrich

Acriflavine-Biotin Conjugate

suitable for fluorescence

Synonym(s):

3,6-Bis(amino)-9-(biotinylamidoethylamino)acridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H31N7O2S
CAS Number:
Molecular Weight:
493.62
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

solubility

DMSO: 1 mg/mL, clear, faintly yellow to yellow

fluorescence

λex 405 nm; λem 484 nm±5 nm in DMSO

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

Nc1ccc2c(NCCNC(=O)CCCC[C@@H]3SC[C@@H]4NC(=O)N[C@H]34)c5ccc(N)cc5nc2c1

InChI

1S/C25H31N7O2S/c26-14-5-7-16-18(11-14)30-19-12-15(27)6-8-17(19)23(16)29-10-9-28-22(33)4-2-1-3-21-24-20(13-35-21)31-25(34)32-24/h5-8,11-12,20-21,24H,1-4,9-10,13,26-27H2,(H,28,33)(H,29,30)(H2,31,32,34)/t20-,21-,24-/m0/s1

InChI key

YWYQMAHVCTUSQN-HFMPRLQTSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruba Khnouf et al.
The Analyst, 139(18), 4564-4571 (2014-07-17)
We have developed a microfluidic device that enhances the sensitivity of protein immunoassays by preconcentrating the protein sample using isotachophoresis (ITP). Two approaches were followed to study the sensitivity gain achieved that way. The first approach was using antibody-coated magnetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service