Skip to Content
Merck
All Photos(1)

Documents

15522

Sigma-Aldrich

N-Boc-D-prolinol

puriss., ≥99.0% (TLC)

Synonym(s):

(R)-1-Boc-2-pyrrolidinemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO3
CAS Number:
Molecular Weight:
201.26
Beilstein:
3650011
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (TLC)

optical activity

[α]20/D +54±2°, c = 5% in methanol

mp

59-62 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCC[C@@H]1CO

InChI

1S/C10H19NO3/c1-10(2,3)14-9(13)11-6-4-5-8(11)7-12/h8,12H,4-7H2,1-3H3/t8-/m1/s1

InChI key

BFFLLBPMZCIGRM-MRVPVSSYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service