Skip to Content
Merck
All Photos(1)

Documents

03819

Sigma-Aldrich

Ethylene glycol monododecyl ether

BioXtra, ≥99.0% (GC)

Synonym(s):

2-(Dodecyloxy)ethanol, C12E1, Dodecylglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OCH2CH2OH
CAS Number:
Molecular Weight:
230.39
Beilstein:
1748129
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

product line

BioXtra

Assay

≥99.0% (GC)

mol wt

230.39 g/mol

refractive index

n20/D 1.446

density

0.873 g/mL at 20 °C (lit.)

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCOCCO

InChI

1S/C14H30O2/c1-2-3-4-5-6-7-8-9-10-11-13-16-14-12-15/h15H,2-14H2,1H3

InChI key

SFNALCNOMXIBKG-UHFFFAOYSA-N

Other Notes

Non-Ionic surfactant. Utilised as a source of C and energy by an organism isolated from activated sludge

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dries Deeren et al.
Acta clinica Belgica, 71(6), 379-382 (2016-05-05)
A recent multicenter study demonstrated that bathing with chlorhexidine reduces the transmission of resistant organisms and the risk of hospital-acquired bloodstream infections in ICUs. We wanted to confirm the feasibility of this strategy in a cohort of patients in a
Y. Ichikawa et al.
J. Ferment. Technol., 56, 403-403 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service