Skip to Content
Merck
All Photos(1)

Key Documents

63139

Sigma-Aldrich

Magnesium sulfate hydrate

purum p.a., ≥99.0% (calc. on dried material, KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · aq
CAS Number:
Molecular Weight:
120.37 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (calc. on dried material, KT)
form:
powder

grade

purum p.a.

Assay

≥99.0% (calc. on dried material, KT)

form

powder

loss

26-36% loss on ignition, 500°C

pH

6.5-8 (25 °C, 120.4 g/L)

anion traces

chloride (Cl-): ≤200 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤500 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byoung Soo Kim et al.
Small (Weinheim an der Bergstrasse, Germany), 15(21), e1900765-e1900765 (2019-04-06)
Various antioxidants are being used to neutralize the harmful effects of reactive oxygen species (ROS) overproduced in diseased tissues and contaminated environments. Polymer-directed crystallization of antioxidants has attracted attention as a way to control drug efficacy through molecular dissolution. However
Eagleson M.
Concise Encyclopedia Chemistry, 377-377 (1994)
Sulfates on Mars: A systematic Raman spectroscopic study of hydration states of magnesium sulfates.
Wang A, et al.
Geochimica et Cosmochimica Acta, 70(24), 6118-6135 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service