Skip to Content
Merck
All Photos(1)

Documents

Y0001929

Tiotropium for system suitability 1

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tiotropium bromide monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22BrNO4S2 · H2O
CAS Number:
Molecular Weight:
490.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tiotropium

manufacturer/tradename

EDQM

format

neat

storage temp.

2-8°C

SMILES string

[H]O[H].O=C(C(C1=C([H])C([H])=C([H])S1)(O[H])C2=C([H])C([H])=C([H])S2)O[C@@]3([H])C([H])([H])[C@@]4([H])[C@@]5([H])[C@@](O5)([H])[C@](C3([H])[H])([H])[N+]4(C([H])([H])[H])C([H])([H])[H].[Br-]

InChI

1S/C19H22NO4S2.BrH.H2O/c1-20(2)12-9-11(10-13(20)17-16(12)24-17)23-18(21)19(22,14-5-3-7-25-14)15-6-4-8-26-15;;/h3-8,11-13,16-17,22H,9-10H2,1-2H3;1H;1H2/q+1;;/p-1/t11?,12-,13+,16?,17?;;

InChI key

MQLXPRBEAHBZTK-YQMLGFMRSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tiotropium for system suitability 1 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Urinary tract,Respiratory system,Gastro-intestinal system,Kidney,Heart

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service