Skip to Content
Merck
All Photos(1)

Key Documents

W292312

Sigma-Aldrich

Propionaldehyde

natural, ≥98%, FG

Synonym(s):

Propanal

Sign Into View Organizational & Contract Pricing

Select a Size

W292312-SAMPLE-K
£45.40
100 G
£86.50
1 KG
£617.00

£45.40


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W292312-SAMPLE-K
£45.40
100 G
£86.50
1 KG
£617.00

About This Item

Linear Formula:
CH3CH2CHO
CAS Number:
Molecular Weight:
58.08
FEMA Number:
2923
Beilstein:
506010
EC Number:
Council of Europe no.:
90
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.002
NACRES:
NA.21
Organoleptic:
chocolate; cocoa; nutty; whiskey; wine-like
grade:
FG
Fragrance grade
Halal
Kosher
natural
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

£45.40


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

vapor density

2 (vs air)

vapor pressure

18.77 psi ( 55 °C)
4.89 psi ( 20 °C)

Assay

≥98%

autoignition temp.

404 °F

expl. lim.

17 %, 26 °F
2.6 %, 31 °F

refractive index

n20/D 1.362 (lit.)

bp

46-50 °C (lit.)

mp

−81 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

chocolate; cocoa; nutty; whiskey; wine-like

storage temp.

2-8°C

SMILES string

[H]C(=O)CC

InChI

1S/C3H6O/c1-2-3-4/h3H,2H2,1H3

InChI key

NBBJYMSMWIIQGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palina S Nepachalovich et al.
Free radical research, 54(10), 732-744 (2020-10-20)
Vicinal diols and its derivatives can be exploited as model compounds for the investigation of radiation-induced free-radical transformations of hydroxyl-containing biomolecules such as carbohydrates, phospholipids, ribonucleotides, amino acids, and peptides. In this paper, for the first time, the prospects of
Li Zhou et al.
The journal of physical chemistry. A, 121(23), 4464-4474 (2017-05-13)
Two different experimental methods (relative rate and absolute rate methods) were used to measure the rate coefficients for the reactions of NO

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service