Skip to Content
Merck
All Photos(1)

Key Documents

T1195

Sigma-Aldrich

Sulforhodamine 101 DHPE

Synonym(s):

Sulforhodamine 101 1,2-dihexadecanoyl-SN-glycero-3-phosphoethanolLamine, triethylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H117N4O14PS2
CAS Number:
Molecular Weight:
1381.84
MDL number:
UNSPSC Code:
12171500
Pricing and availability is not currently available.

shipped in

dry ice

storage temp.

−20°C

Application

Energy transfer acceptor used in resonance energy transfer microscopy.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service