Skip to Content
Merck
All Photos(1)

Key Documents

SIAL311NZ0.5C

Microcentrifuge Tube, 0.5 mL

conical bottom, natural

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

conical bottom
natural
polypropylene

packaging

pkg of 10 × 1000 ea

H

31.6 mm

capacity

0.5 mL

diam.

7.7 mm

Looking for similar products? Visit Product Comparison Guide

General description

0.5 mL Microcentrifuge tubes with attached click lock lids. Available in a variety of colours.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service