Skip to Content
Merck
All Photos(1)

Documents

QCMIC213

Millipore

Salmonella - Sludge QC 4 Levels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

grade

certified reference material

packaging

set of

manufacturer/tradename

RTC QC-MIC-213

application(s)

environmental
food and beverages

shipped in

wet ice

storage temp.

2-8°C

suitability

Salmonella spp.

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Packaging

Set contains 8 sterile screw cap vials each containing a Vitroid (a small disk containing the possible organisms) and 4 jars of sludge.
Contains not the phosphate buffered hydrolyzing solution. The buffer can be ordered separately under the number Z699489 (Aldrich).

Other Notes

Refer to the Certificate of Analysis for additional information.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service