Skip to Content
Merck
All Photos(1)

Documents

QC1117

Supelco

pH 10 Buffer

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 100 mL

manufacturer/tradename

RTC QCI-027-10-.100

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

These products are discontinued but the list below shows products that can be used as replacement.

Obsolete Part NumberReplacement Part Number
QC1117-250 ML1094090500
QC1117-500 ML1094090500
QC1117-1L1094091000
QC1117-10 L1094389010

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service