PHR8904
Desfluoromethyl fluticasone propionate disulfide
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
17,17′-(Disulphanediyldicarbonyl)bis(6α,9-difluoro-11β-hydroxy-16α-methyl-3-oxoandrosta-1,4-dien-17α-yl) dipropanoate
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
API family
fluticasone
form
solid
packaging
pkg of 100 mg
application(s)
pharmaceutical small molecule
General description
Application
Analysis Note
Other Notes
Footnote
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service