Skip to Content
Merck
All Photos(1)

Documents

PHR3826

Supelco

Triethylene Glycol

certified reference material, pharmaceutical secondary standard

Synonym(s):

Triethylene glycol, Triglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)2CH2CH2OH
CAS Number:
Molecular Weight:
150.17
Beilstein:
969357
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1683708

vapor density

5.2 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

699 °F

expl. lim.

9.2 %

packaging

pkg of 1 mL

refractive index

n20/D 1.455 (lit.)

bp

125-127 °C/0.1 mmHg (lit.)

mp

−7 °C (lit.)

density

1.124 g/mL at 20 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

OCCOCCOCCO

InChI

1S/C6H14O4/c7-1-3-9-5-6-10-4-2-8/h7-8H,1-6H2

InChI key

ZIBGPFATKBEMQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

330.8 °F

Flash Point(C)

166 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service