Skip to Content
Merck
All Photos(1)

Key Documents

PHR3567

Supelco

Moexipril hydrochloride

certified reference material, pharmaceutical secondary standard

Synonym(s):

(3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

PHR3567-200MG
£225.00

£225.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
PHR3567-200MG
£225.00

About This Item

Empirical Formula (Hill Notation):
C27H35N2O7Cl
CAS Number:
Molecular Weight:
535.03
MDL number:
UNSPSC Code:
41116107

£225.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1445426

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

application(s)

pharmaceutical

SMILES string

Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2Cc3cc(OC)c(OC)cc3C[C@H]2C(O)=O

InChI

1S/C27H34N2O7.ClH/c1-5-36-27(33)21(12-11-18-9-7-6-8-10-18)28-17(2)25(30)29-16-20-15-24(35-4)23(34-3)14-19(20)13-22(29)26(31)32;/h6-10,14-15,17,21-22,28H,5,11-13,16H2,1-4H3,(H,31,32);1H/t17-,21-,22-;/m0./s1

InChI key

JXRAXHBVZQZSIC-JKVLGAQCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Biochem/physiol Actions

ACE-Inhibitor; Antihypertensive
Moexipril is an ACE-Inhibitor; Antihypertensive; Unlike other ACE inhibitors, prolongs QT interval leading to torsades de pointes and cardiad arrhythmia;

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959(changes by product) in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service