Skip to Content
Merck
All Photos(1)

Key Documents

PHR1695

Supelco

Phenylephrine Related Compound C

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-(3-Hydroxyphenyl)-2-(methylamino)ethanone hydrochloride, Phenylephrone hydrochloride, 3′-Hydroxyphenyl-2-methylaminoacetophenone hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
£598.00

£598.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
£598.00

About This Item

Empirical Formula (Hill Notation):
C9H11NO2 · HCl
CAS Number:
Molecular Weight:
201.65
UNSPSC Code:
41116107
NACRES:
NA.24

£598.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1533024

API family

phenylephrine

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C9H11NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,10-11H,6H2,1H3;1H

InChI key

DEOGEZWYKPQJLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

It is an impurity of the widely used decongestant drug― phenylephrine, a selective α1-adrenergic receptor agonist.

Application

This pharmaceutical secondary standard can also be used as follows:

  • Impurity profiling of diphenhydramine and phenylephrine hydrochloride oral solutions using ultra-high performance liquid chromatography (UHPLC)

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2623 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Xiao et al.
Journal of cellular biochemistry, 118(11), 3899-3910 (2017-04-09)
Lack of effective anti-cardiac hypertrophy drugs creates a major cause for the increasing prevalence of heart failure. In the present study, we determined the anti-hypertrophy and anti-fibrosis potential of a natural plant triterpenoid, Cucurbitacin B both in vitro and in
Qing-Qing Wu et al.
British journal of pharmacology, 175(9), 1548-1566 (2018-02-16)
Aucubin, the predominant component of Eucommia ulmoides Oliv., has been shown to have profound effects on oxidative stress. As oxidative stress has previously been demonstrated to contribute to acute and chronic myocardial injury, we tested the effects of aucubin on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service