N4909
2-(2,3-Naphthalimino)ethyl trifluoromethanesulfonate
Synonym(s):
Methanesulfonic acid, trifluoro-2-(1,3-dihydro-1,3-dioxo-2H-benz[F]iso-indol- 2-yl)ethyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H10F3NO5S
CAS Number:
Molecular Weight:
373.30
MDL number:
UNSPSC Code:
12171500
Recommended Products
mol wt
373.3
shipped in
dry ice
storage temp.
−20°C
Application
Fluorescent reagent that reacts rapidly with anions of carboxylic acids in acetonitrile, carnitines, and acylcarnitines in tissue for sensitive fluorimetric detection.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service