K4512
Kavergal 1
analytical standard, 65% (NMR), solid
Synonym(s):
[(+)-Catechin-(4α→8)-(+)-gallocatechin-(4α→8)]n-(+)-catechin
About This Item
grade
analytical standard
Quality Level
Assay
65% (NMR)
form
solid
mol wt
average mol wt 8,000 Da (n = 1 to 10)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
solubility
ethanol: 40%
H2O: soluble
format
neat
storage temp.
2-8°C
Application
Biochem/physiol Actions
Caution
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service