Skip to Content
Merck
All Photos(1)

Documents

F0189900

Flunarizine dihydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-[Bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)piperazine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26F2N2 · 2HCl
CAS Number:
Molecular Weight:
477.42
Beilstein:
4284243
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

flunarizine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Fc1ccc(cc1)C(N2CCN(CC2)C\C=C\c3ccccc3)c4ccc(F)cc4

InChI

1S/C26H26F2N2.2ClH/c27-24-12-8-22(9-13-24)26(23-10-14-25(28)15-11-23)30-19-17-29(18-20-30)16-4-7-21-5-2-1-3-6-21;;/h1-15,26H,16-20H2;2*1H/b7-4+;;

InChI key

RXKMOPXNWTYEHI-RDRKJGRWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flunarizine dihydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Blocks T-type Ca2+/Na+ channels; inhibits K+-induced catecholamine release from chromaffin cells

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grażyna Biała et al.
Pharmacological reports : PR, 66(2), 212-222 (2014-06-10)
Nonsomatic signs of psychostimulant withdrawal, difficult to demonstrate in animal paradigms, may appear to promote drug seeking and drug relapse in humans; thus, it is important to understand the mechanisms that mediate this kind of behaviors. The present study was
Wen Liu et al.
Endocrinology, 155(5), 1738-1750 (2014-02-13)
The insect steroid hormone 20-hydroxyecdysone (20E) regulates gene transcription via a genomic pathway by forming a transcription complex that binds to DNA with the help of the chaperone proteins, heat shock proteins (Hsps) Hsc70 and Hsp90. However, the nongenomic mechanisms
Jonathan W Astin et al.
Molecular cancer therapeutics, 13(10), 2450-2462 (2014-07-24)
The growth of new lymphatic vessels (lymphangiogenesis) in tumors is an integral step in the metastatic spread of tumor cells, first to the sentinel lymph nodes that surround the tumor and then elsewhere in the body. Currently, no selective agents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service