Skip to Content
Merck
All Photos(1)

Documents

DLW354233

Sigma-Aldrich

Corning® Collagen IV Mouse Natural 1 mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

mouse (Engelbreth-Holm-Swarm (EHS) lathrytic mouse tumor)

Assay

≥90% (SDS-PAGE)

form

frozen liquid (Frozen in 0.05 M HCI)

packaging

pkg of 1 mg

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Col4a4(12829)

General description

Corning® Collagen IV mouse 1mg, is used as a thin coating on tissue-culture surfaces to promote cell attachment and proliferation and to study the collagen′s effects on cell behavior.

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service