Skip to Content
Merck
All Photos(1)

Key Documents

C9651

Sigma-Aldrich

Ammonium cerium(IV) sulfate dihydrate

≥95%

Synonym(s):

Ceric ammonium sulfate, Cerium(IV) ammonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)4(SO4)4 · 2H2O
CAS Number:
Molecular Weight:
632.55
MDL number:
UNSPSC Code:
12352300

Assay

≥95%

mp

130 °C (lit.)

SMILES string

N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Ce.4H3N.4H2O4S.2H2O/c;;;;;4*1-5(2,3)4;;/h;4*1H3;4*(H2,1,2,3,4);2*1H2/q+4;;;;;;;;;;/p-4

InChI key

VCNAMBGKEDPVGQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kaul et al.
Amino acids, 34(2), 315-320 (2006-11-07)
An assessment of the potential of proline to scavenge free radicals was made in a couple of in vitro assay systems, namely graft co-polymerization and autooxidation of pyrogallol. Both these assays are essentially dependent upon free radical mechanisms. Graft co-polymerization
K H Gordon et al.
Organic letters, 3(1), 53-56 (2001-06-30)
[figure: see text] A novel benzyloxyaniline linker that uses ceric ammonium nitrate (CAN) as a cleavage reagent is described. Its application in the solid-phase synthesis of N-unsubstituted beta-lactams and secondary amides furnishes compounds in moderate to excellent yield (45-91%) and
K Tsuda et al.
Clinical chemistry, 41(4), 581-585 (1995-04-01)
We have modified an automated measurement system of urinary iodine (UI) and established a sensitive UI assay system by using ultraviolet (UV) digestion. The automated system is sensitive enough to detect concentrations of UI < 0.78 mumol/L (< 10 micrograms/dL)
Effect of high room temperature on urinary iodine assay.
N T Dung et al.
Clinical chemistry, 43(6 Pt 1), 1084-1085 (1997-06-01)
Babu G Gowda et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 18(6), 671-674 (2002-06-27)
Two simple and sensitive indirect spectrophotometric methods for the assay of propranolol hydrochloride (PPH) and piroxicam (PX) in pure and pharmaceutical formulations have been proposed. The methods are based on the oxidation of PPH by a known excess of standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service