Skip to Content
Merck
All Photos(1)

Documents

BP141

Ergotamine tartrate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Ergotamine D-tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H35N5O5 · 0.5C4H6O6
CAS Number:
Molecular Weight:
656.70
Beilstein:
6173437
MDL number:
UNSPSC Code:
41116107

grade

pharmaceutical primary standard

manufacturer/tradename

BP

mp

~195 °C (dec.) (lit.)

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1C[C@@H](C=C2C1Cc3c[nH]c4cccc2c34)C(=O)N[C@]5(C)O[C@@]6(O)C7CCCN7C(=O)[C@H](Cc8ccccc8)N6C5=O.CN9C[C@@H](C=C%10C9Cc%11c[nH]c%12cccc%10c%11%12)C(=O)N[C@]%13(C)O[C@@]%14(O)C%15CCCN%15C(=O)[C@H](Cc%16ccccc%16)N%14C%13=O

InChI

1S/2C33H35N5O5.C4H6O6/c2*1-32(35-29(39)21-15-23-22-10-6-11-24-28(22)20(17-34-24)16-25(23)36(2)18-21)31(41)38-26(14-19-8-4-3-5-9-19)30(40)37-13-7-12-27(37)33(38,42)43-32;5-1(3(7)8)2(6)4(9)10/h2*3-6,8-11,15,17,21,25-27,34,42H,7,12-14,16,18H2,1-2H3,(H,35,39);1-2,5-6H,(H,7,8)(H,9,10)/t2*21-,25-,26+,27+,32-,33+;1-,2-/m111/s1

InChI key

CJMJLDQKTOJACI-BGQAIRJTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 200 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Repr. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service