Skip to Content
Merck
All Photos(1)

Key Documents

94120

Sigma-Aldrich

Methyl undecanoate

≥98.0% (GC)

Synonym(s):

Undecanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CO2CH3
CAS Number:
Molecular Weight:
200.32
Beilstein:
1762129
EC Number:
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

Assay

≥98.0% (GC)

refractive index

n20/D 1.429 (lit.)
n20/D 1.429

density

0.872 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCC(=O)OC

InChI

1S/C12H24O2/c1-3-4-5-6-7-8-9-10-11-12(13)14-2/h3-11H2,1-2H3

InChI key

XPQPWPZFBULGKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service