Skip to Content
Merck
All Photos(1)

Key Documents

90913

Supelco

7-[3-(Dansylamido)propyl]-1-oxa-4,10-dithia-7-azacyclododecane

Selectophore, ≥97.0%

Synonym(s):

N-[3-(1-Oxa-4,10-dithia-7-aza-7-cyclododecyl)propyl]dansylamide, [12]aneNS2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H35N3O3S3
CAS Number:
Molecular Weight:
497.74
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Assay

≥97.0% (HPLC)
≥97.0%

λmax

251 nm±5 nm in acetonitrile: water (4:1)

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)NCCCN3CCSCCOCCSCC3

General description

Visit our Sensor Applications portal to learn more.

Application

Selective fluoroionophore for Hg(II), measuring range 1x10-4 - 5x10-12 mol/L.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shamsipur M., et al.
Analytica Chimica Acta, 533, 17-24 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service