Skip to Content
Merck
All Photos(1)

Key Documents

89998

Supelco

10-[3-(Dansylamido)propyl]-1,4,7-trithia-10-azacyclododecane

Selectophore, ≥95.0%

Synonym(s):

N-[3-(1,4,7-Trithia-10-aza-10-cyclododecyl)propyl]dansylamide, [12]aneNS3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H35N3O2S4
CAS Number:
Molecular Weight:
513.80
Beilstein:
10212902
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Assay

≥95.0%

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)NCCCN3CCSCCSCCSCC3

General description

Visit our Sensor Applications portal to learn more.

Application

Selective fluoroionophore for Ag+, measuring range 1.7x10-2 - 5.0x10-7 mol/L.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shamsipur, M., et al.
Sensors and Actuators B, Chemical, 113, 892-899 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service