Skip to Content
Merck
All Photos(1)

Key Documents

89838

Supelco

Sodium tetra(p-tolyl)borate

Selectophore, for ion-selective electrodes, ≥99.0%

Synonym(s):

Sodium tetrakis(4-methylphenyl)borate, Tetra(p-tolyl)boron sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28BNa
CAS Number:
Molecular Weight:
398.32
Beilstein:
4072242
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.0% (NT)
≥99.0%

form

solid

SMILES string

[Na+].Cc1ccc(cc1)[B-](c2ccc(C)cc2)(c3ccc(C)cc3)c4ccc(C)cc4

InChI

1S/C28H28B.Na/c1-21-5-13-25(14-6-21)29(26-15-7-22(2)8-16-26,27-17-9-23(3)10-18-27)28-19-11-24(4)12-20-28;/h5-20H,1-4H3;/q-1;+1

InChI key

VVPGHQQFKGSIPJ-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service