Skip to Content
Merck
All Photos(1)

Key Documents

89536

Sigma-Aldrich

Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] oxalate

≥90% (HPLC)

Synonym(s):

Dipentyl O,O′-oxalyl-bis(3,5,6-trichlorosalicylate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24Cl6O8
CAS Number:
Molecular Weight:
677.18
Beilstein:
2932173
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥90% (HPLC)

SMILES string

CCCCCOC(=O)c1c(Cl)c(Cl)cc(Cl)c1OC(=O)C(=O)Oc2c(Cl)cc(Cl)c(Cl)c2C(=O)OCCCCC

Other Notes

Reagent for the generation of chemiluminescence with H2O2. Used for the determination of fluorescent compounds[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Yoshinaga et al.
Bulletin of the Chemical Society of Japan, 74, 1507-1507 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service