Skip to Content
Merck
All Photos(1)

Key Documents

89262

Sigma-Aldrich

1,1,2-Trimethyl-3-(4-sulfobutyl)benz[e]indolium, inner salt

purum, ≥98.0% (HPLC)

Synonym(s):

1,1,2-Trimethyl-3-(4-sulfobutyl)-1H-benz[e]indolium hydroxide, inner salt, 1-(4-Sulfobutyl)-2,3,3-trimethyl-4,5-benzindolium, inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO3S
CAS Number:
Molecular Weight:
345.46
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

purum

Assay

≥98.0% (HPLC)

mp

274-278 °C

fluorescence

λex 319 nm; λem 457 nm in DMSO

SMILES string

CC1=[N+](CCCCS([O-])(=O)=O)c2ccc3ccccc3c2C1(C)C

InChI

1S/C19H23NO3S/c1-14-19(2,3)18-16-9-5-4-8-15(16)10-11-17(18)20(14)12-6-7-13-24(21,22)23/h4-5,8-11H,6-7,12-13H2,1-3H3

InChI key

GJCURKVOCXJAIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service