89172
Bis(2,2,2-trichloroethyl) phosphorochloridate
purum, ≥98.0% (AT)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(Cl3CCH2O)2P(O)Cl
CAS Number:
Molecular Weight:
379.22
Beilstein:
1820707
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
purum
Assay
≥98.0% (AT)
mp
45-47 °C (lit.)
SMILES string
ClC(Cl)(Cl)COP(Cl)(=O)OCC(Cl)(Cl)Cl
InChI
1S/C4H4Cl7O3P/c5-3(6,7)1-13-15(11,12)14-2-4(8,9)10/h1-2H2
InChI key
ZHHCWQGVXYGWCW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Building block to introduce a (2,2,2-trichloro)ethyl protected phosphate in tyrosine; for protected phosphoserine
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Paquet et al.
The journal of peptide research : official journal of the American Peptide Society, 50(4), 262-268 (1997-11-14)
Suitability of bis-(2,2,2-trichloro)ethyl (Tc) groups for protection of phosphate moiety in Boc-mode synthesis of phosphotyrosine peptides is demonstrated Boc-Tyr(PO3Tc2)-OH and Fmoc-Tyr(PO3Tc2)-OH were prepared by acylating H-Tyr(PO3Tc2)-OH with (Boc)2O and Fmoc-ONSu, respectively. Phosphorus introduction was achieved by phosphorylating Boc-Tyr-OBzl with Tc
A Paquet
International journal of peptide and protein research, 39(1), 82-86 (1992-01-01)
Boc-Ser(PO3Tc2)-OH, Z-Ser(PO3Tc2)-OH and Fmoc-Ser(PO3Tc2)-OH, derivatives useful for peptide synthesis, have been obtained in high yields by acylation of H-Ser(PO3Tc2)-OH.CF3COOH. The latter was obtained from Boc- or Z-Ser(PO3Tc2)-OBzl by simultaneous removal of the amino- and carboxy-protecting groups by Pd-catalyzed hydrogenolysis in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service