Skip to Content
Merck
All Photos(2)

Documents

87204

Supelco

1-Phenyltetradecane

analytical standard

Synonym(s):

Tetradecylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)13CH3
CAS Number:
Molecular Weight:
274.48
Beilstein:
2047457
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.481

bp

359 °C (lit.)

mp

16 °C (lit.)

density

0.854 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCCCCCCCCCCCCCc1ccccc1

InChI

1S/C20H34/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-20-18-15-13-16-19-20/h13,15-16,18-19H,2-12,14,17H2,1H3

InChI key

JZALLXAUNPOCEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Phenyltetradecane is an organic compound and an environmental contaminant belonging to the class of alkylbenzenes.

Application

1-Phenyltetradecane may be used as an analytical reference standard for the quantification of the analyte in environmental samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of long-chain alkylbenzenes in environmental samples by argentation thin-layer chromatography/high-resolution gas chromatography and gas chromatography/mass spectrometry
Eganhouse.PR, et al.
Analytical Chemistry, 55(13), 2120-2126 (1983)
Detection of organic contamination in sediments by double-shoot pyrolysis-GC/MS
Teran.A, et al.
Environmental Chemistry Letters, 7(4), 301-308 (2009)
Csanád Rédei et al.
Journal of chromatography. A, 1603, 348-354 (2019-06-06)
Uncommon retention behavior of a series of n-alkylbenzene homologues as well as the effect of different sample solvents on chromatographic efficiency were studied in supercritical fluid chromatography. After testing various columns, an alkylamide stationary phase was selected for detailed studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service