Skip to Content
Merck
All Photos(1)

Key Documents

86335

Sigma-Aldrich

Sodium taurochenodeoxycholate

≥97.0% (TLC)

Synonym(s):

2-([3α,7α-Dihydroxy-24-oxo-5β-cholan-24-yl]amino)ethanesulfonic acid sodium salt, 3α,7α-Dihydroxy-5β-cholan-24-oic acid N-(2-sulfoethyl)amide sodium salt, Taurochenodeoxycholic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44NNaO6S
CAS Number:
Molecular Weight:
521.69
Beilstein:
3901205
MDL number:
UNSPSC Code:
12352200

description

anionic

Assay

≥97.0% (TLC)

impurities

≤7% water

solubility

H2O: 0.1 g/mL, clear, faintly yellow

SMILES string

[H][C@@]12[C@]([C@](CC[C@@H](O)C3)(C)[C@]3([H])C[C@H]2O)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC(NCCS(=O)([O-])=O)=O.[Na+]

InChI

1S/C26H45NO6S.Na/c1-16(4-7-23(30)27-12-13-34(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29;/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);/q;+1/p-1/t16-,17+,18-,19-,20+,21+,22-,24?,25+,26-;/m1./s1

InChI key

IYPNVUSIMGAJFC-ULWOGUKGSA-M

Looking for similar products? Visit Product Comparison Guide

General description

An ionic detergent used for solubilizing lipids and proteins.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susanne Naumann et al.
Nutrients, 11(9) (2019-09-08)
Interference of dietary fibres with the enterohepatic circulation of bile acids is proposed as a mechanism for lowering cholesterol. We investigated how lupin hull and cotyledon dietary fibres interact with primary bile acids using an in vitro model under simulated
Susanne Naumann et al.
Nutrients, 11(6) (2019-06-28)
Dietary fibres are reported to interact with bile acids, preventing their reabsorption and promoting their excretion into the colon. We used a method based on in vitro digestion, dialysis, and kinetic analysis to investigate how dietary fibre enriched food ingredients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service