Skip to Content
Merck
All Photos(1)

Key Documents

85897

Sigma-Aldrich

Strontium acetate hydrate

puriss. p.a., ≥99.995% (calc. based on trace metal analysis)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Sr · xH2O
CAS Number:
Molecular Weight:
205.71 (anhydrous basis)
Beilstein:
3692534
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

grade

puriss. p.a.

Assay

≥99.995% (calc. based on trace metal analysis)

concentration

40.63-40.80% Sr

impurities

≤0.005% total of impurities (ICP-OES)
≤5.0% water

cation traces

Ca: ≤1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤10 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.CC(=O)O[Sr]OC(C)=O

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service