Skip to Content
Merck
All Photos(1)

Key Documents

84740

Sigma-Aldrich

Sulfur trioxide trimethylamine complex

technical, ≥90% (S)

Synonym(s):

NSC 9838, Trimethylamine sulfur trioxide complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N · SO3
CAS Number:
Molecular Weight:
139.17
Beilstein:
3681759
EC Number:
MDL number:
UNSPSC Code:
12352000

grade

technical

Assay

≥90% (S)

reaction suitability

reagent type: oxidant

mp

232 °C (dec.) (lit.)

SMILES string

CN(C)C.O=S(=O)=O

InChI

1S/C3H9N.O3S/c2*1-4(2)3/h1-3H3;

InChI key

DXASQZJWWGZNSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for sulfation reactions involving:
  • Synthesis of sulfate-conjugated resveratrol metabolites
  • Chitooligosaccharides used for anti-HIV-1 activity
  • Dodecyl thioglycopyranoside used as a surfactant for enantiomeric separation
  • Glycosaminoglycans which facilitate activation of signaling pathways dependent on sulfation pattern
  • Polysaccharides as heparan sulfate mimetics

Nucleophile for the synthesis of α-tosyloxy ketones

Other Notes

Reagent for the sulfation of alcohols

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Guilbert et al.
Tetrahedron Letters, 35, 6563-6563 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service