Skip to Content
Merck
All Photos(1)

Documents

78814

Sigma-Aldrich

Ethyl 3-hydroxy-4-methoxy-mandelate

≥99.0%

Synonym(s):

3-Hydroxy-4-methoxy-mandelic acid-ethyl ester, Ethyl hydroxy-(3-hydroxy-4-methoxy-phenyl)-acetate, Isovanilmandelic acid-ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O5
CAS Number:
Molecular Weight:
226.23
Beilstein:
3137338
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0%

form

powder

mp

124-130 °C

SMILES string

CCOC(=O)C(O)c1ccc(OC)c(O)c1

InChI

1S/C11H14O5/c1-3-16-11(14)10(13)7-4-5-9(15-2)8(12)6-7/h4-6,10,12-13H,3H2,1-2H3

InChI key

TVPGKWGQPSIRLU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service