Skip to Content
Merck
All Photos(1)

Key Documents

72593

Supelco

Nitrite ionophore II

Selectophore

Synonym(s):

Dicyanocobyrinic acid heptapropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C68H101CoN6O14
CAS Number:
Molecular Weight:
1285.49
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

form

liquid

storage temp.

2-8°C

SMILES string

CCCOC(=O)CC[C@@H]1C2=CC3=NC([C@@H](CCC(=O)OCCC)C3(C)C)=C(C)C4=NC([C@H](CC(=O)OCCC)[C@@]4(C)CCC(=O)OCCC)[C@]5(C)N=C([C@@H](CCC(=O)OCCC)[C@]5(C)CC(=O)OCCC)C(C)=C(N2[Co](C#N)C#N)[C@@]1(C)CC(=O)OCCC

InChI

1S/C66H101N4O14.2CN.Co/c1-16-31-78-50(71)26-23-44-48-39-49-62(10,11)45(24-27-51(72)79-32-17-2)57(68-49)42(8)59-63(12,30-29-53(74)81-34-19-4)47(38-54(75)82-35-20-5)61(69-59)66(15)65(14,41-56(77)84-37-22-7)46(25-28-52(73)80-33-18-3)58(70-66)43(9)60(67-48)64(44,13)40-55(76)83-36-21-6;2*1-2;/h39,44-47,61H,16-38,40-41H2,1-15H3;;;/q-1;;;+1/t44-,45-,46-,47+,61?,63-,64+,65+,66+;;;/m1.../s1

InChI key

ISWKZOZLNIEXOZ-GRQAXCPQSA-N

General description

Visit our Sensor Applications portal to learn more.

Physical form

solution in 0.5 mL tetrahydrofuran

Other Notes

Ionophore used for the determination of nitrogen oxide gas

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.A. O'Reilly et al.
Analytical Chemistry, 63, 1278-1278 (1991)
Alicia Bellomo et al.
Immunity, 53(1), 127-142 (2020-06-21)
Located within red pulp cords, splenic red pulp macrophages (RPMs) are constantly exposed to the blood flow, clearing senescent red blood cells (RBCs) and recycling iron from hemoglobin. Here, we studied the mechanisms underlying RPM homeostasis, focusing on the involvement
Polymer membrane-based biosensors using vitamin B12 derivatives.
S Daunert et al.
Progress in clinical and biological research, 292, 215-225 (1989-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service