Skip to Content
Merck
All Photos(1)

Key Documents

72195

Sigma-Aldrich

Netropsin dihydrochloride hydrate

≥96.0% (HPCE)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26N10O3 · 2HCl · xH2O
CAS Number:
Molecular Weight:
503.39 (anhydrous basis)
Beilstein:
3852617
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

Assay

≥96.0% (HPCE)

shelf life

limited shelf life, expiry date on the label

impurities

10% water

antibiotic activity spectrum

viruses

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

2-8°C

SMILES string

Cl.Cl.Cn1cc(NC(=O)c2cc(NC(=O)CNC(N)=N)cn2C)c(c1)C(=O)NCCC(N)=N

InChI

1S/C18H26N10O3.2ClH/c1-27-8-11(16(30)23-4-3-14(19)20)12(9-27)26-17(31)13-5-10(7-28(13)2)25-15(29)6-24-18(21)22;;/h5,7-9H,3-4,6H2,1-2H3,(H3,19,20)(H,23,30)(H,25,29)(H,26,31)(H4,21,22,24);2*1H

InChI key

AGIFDNIQJFEFMQ-UHFFFAOYSA-N

Other Notes

Non-intercalating DNA binding agent with preference for A+T-rich regions

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Portugal et al.
European journal of biochemistry, 167(2), 281-289 (1987-09-01)
Techniques of DNase I and micrococcal nuclease footprinting have been used to compare the binding sites for berenil, netropsin and distamycin on two different DNA fragments. Each ligand binds to the A + T-rich zones which contain clusters of at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service