Skip to Content
Merck
All Photos(1)

Documents

70930

Sigma-Aldrich

1-Naphthaleneacetic anhydride

purum, for fluorescence, ≥97.0% (T)

Synonym(s):

1-Naphthylacetic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H7CH2CO)2O
CAS Number:
Molecular Weight:
354.40
Beilstein:
2168120
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

for fluorescence
purum

Assay

≥97.0% (T)

mp

116-119 °C (lit.)
116-119 °C

storage temp.

2-8°C

SMILES string

O=C(Cc1cccc2ccccc12)OC(=O)Cc3cccc4ccccc34

InChI

1S/C24H18O3/c25-23(15-19-11-5-9-17-7-1-3-13-21(17)19)27-24(26)16-20-12-6-10-18-8-2-4-14-22(18)20/h1-14H,15-16H2

InChI key

ACONQQKANQRXQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Derivatizing agent for the determination of airborne polyamines with HPLC, with N,N-Diisopropyl-naphthylacetamide as standard

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W.S. Wu, V.S. Gaind
Journal of Liquid Chromatography, 15, 267-267 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service