Skip to Content
Merck
All Photos(1)

Key Documents

70700

Sigma-Aldrich

(±)-1-(2-Naphthyl)ethanol

puriss., ≥99.0% (HPLC)

Synonym(s):

(±)-α-Methyl-2-naphthalenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12O
CAS Number:
Molecular Weight:
172.22
Beilstein:
1907449
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (HPLC)

mp

71-73 °C

SMILES string

CC(O)c1ccc2ccccc2c1

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service