Skip to Content
Merck
All Photos(1)

Documents

64994

Sigma-Aldrich

(Z)-(Methoxyimino)[2-(tritylamino)thiazol-4-yl]acetic acid monohydrochloride

purum, ≥95.0% (HPLC)

Synonym(s):

(Z)-[2-(Tritylamino)thiazol-4-yl](methoxyimino)acetic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N3O3S · HCl
CAS Number:
Molecular Weight:
479.98
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥95.0% (HPLC)

SMILES string

Cl.CO\N=C(/C(O)=O)c1csc(NC(c2ccccc2)(c3ccccc3)c4ccccc4)n1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service