Skip to Content
Merck
All Photos(1)

Key Documents

53956

Sigma-Aldrich

Hydroquinine 2,5-diphenyl-4,6-pyrimidinediyl diether

purum, ≥97.0% (sum of enantiomers, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H60N6O4
CAS Number:
Molecular Weight:
881.11
MDL number:
UNSPSC Code:
12352000
Pricing and availability is not currently available.

grade

purum

Assay

≥97.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D +460±5°, c = 1.2% in methanol

mp

245-248 °C (lit.)
245-248 °C

SMILES string

CC[C@H]1CN2CCC1C[C@@H]2[C@H](Oc3nc(nc(O[C@@H]([C@@H]4CC5CC[N@H]4C[C@H]5CC)c6ccnc7ccc(OC)cc67)c3-c8ccccc8)-c9ccccc9)c%10ccnc%11ccc(OC)cc%10%11

InChI

1S/C56H60N6O4/c1-5-35-33-61-27-23-39(35)29-49(61)52(43-21-25-57-47-19-17-41(63-3)31-45(43)47)65-55-51(37-13-9-7-10-14-37)56(60-54(59-55)38-15-11-8-12-16-38)66-53(50-30-40-24-28-62(50)34-36(40)6-2)44-22-26-58-48-20-18-42(64-4)32-46(44)48/h7-22,25-26,31-32,35-36,39-40,49-50,52-53H,5-6,23-24,27-30,33-34H2,1-4H3/t35-,36?,39?,40+,49?,50?,52+,53+/m0/s1

InChI key

SWKRDCRSJPRVNF-RMMANSCUSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Ligand for the asymmetric dihydroxylation of olefins. It is superior to other ligands for the AD of monosubstituted terminal olefins[1][2]

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.A. Crispino et al.
The Journal of Organic Chemistry, 58, 3785-3785 (1993)
H.C. Kolb et al.
Chemical Reviews, 94, 2483-2483 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service