Skip to Content
Merck
All Photos(2)

Key Documents

49558

Supelco

2,6-Dimethyl-4-quinolinecarboxylic acid N-hydroxysuccinimide ester

for HPLC derivatization, LiChropur, ≥98.0% (HPLC)

Synonym(s):

N-Succinimidyl-2,6-dimethyl-4-quinolinecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O4
CAS Number:
Molecular Weight:
298.29
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥98.0% (HPLC)

quality

LiChropur

technique(s)

HPLC: suitable

storage temp.

2-8°C

SMILES string

Cc1ccc2nc(C)cc(C(=O)ON3C(=O)CCC3=O)c2c1

InChI

1S/C16H14N2O4/c1-9-3-4-13-11(7-9)12(8-10(2)17-13)16(21)22-18-14(19)5-6-15(18)20/h3-4,7-8H,5-6H2,1-2H3

InChI key

HDQNGUNXGKGYSR-UHFFFAOYSA-N

General description

Fluorescent probe DMQCOSu for analysis of aliphatic primary amines and diamines in water (tap water and river water), human urine and sera without solvent extraction. The detection limit in real samples is between 0.07–4.00 nmol/L.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC Analysis of Aliphatic Primary Amines and Diamines After Derivatization with 2,6-Dimethyl-4-quinolinecarboxylic Acid N-Hydroxysuccinimide Ester
Huang, K., et al.
Chromatographia, 70, 747-752 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service