Skip to Content
Merck
All Photos(1)

Key Documents

47725

Sigma-Aldrich

1-Formylpiperidine

purum, ≥99.0% (GC)

Synonym(s):

N-Carbaldehyde piperidine, NFP, Piperidine-1-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
107697
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor pressure

0.1 mmHg ( 25 °C)

grade

purum

Assay

≥99.0% (GC)

refractive index

n20/D 1.484 (lit.)
n20/D 1.485

bp

222 °C (lit.)

density

1.019 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)N1CCCCC1

InChI

1S/C6H11NO/c8-6-7-4-2-1-3-5-7/h6H,1-5H2

InChI key

FEWLNYSYJNLUOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Direct amidation of azoles with formamides via metal-free C-H activation in the presence of tert-butyl perbenzoate
  • Vilsmeier-type reactions of pyrazoles with amides in the presence of phosphorous oxychloride
  • CO-free aminocarbonylation of N-substituted formamides with aryl iodides/bromides catalyzed by palladium acetate and Xantphos for synthesis of arylamides
  • Synthesis of alternating copolymers for organic photovoltaic applications
  • One-pot double functionalization of π-deficient heterocyclic lithium reagents
  • Synthesis of thermally stable piezofluorochromic aggregation-induced emission compounds

Other Notes

Used in formyl transfer to Grignard reagents

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G.A. Olah et al.
Organic Syntheses, 64, 114-114 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service