Skip to Content
Merck
All Photos(1)

Key Documents

45509

Supelco

Flurenol-butyl

PESTANAL®, analytical standard

Synonym(s):

Butyl 9-hydroxyfluorene-9-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O3
CAS Number:
Molecular Weight:
282.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

product line

PESTANAL®

application(s)

agriculture
environmental

SMILES string

CCCCOC(=O)C1(O)c2ccccc2-c3ccccc13

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service