Skip to Content
Merck
All Photos(1)

Key Documents

44912

Sigma-Aldrich

9-Isothiocyanato-10-methylacridinium triflate

for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11F3N2O3S2
CAS Number:
Molecular Weight:
400.40
Beilstein:
6472816
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

fluorescence

λex 410 nm; λem 450 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

SMILES string

[O-]S(=O)(=O)C(F)(F)F.C[n+]1c2ccccc2c(N=C=S)c3ccccc13

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

In addition to the emission maximum at 450 nm, there are lower maxima at 467 nm and 500 nm.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service