Skip to Content
Merck
All Photos(1)

Key Documents

44197

Supelco

2-Dodecylcyclobutanone

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£122.00

£122.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
£122.00

About This Item

Empirical Formula (Hill Notation):
C16H30O
CAS Number:
Molecular Weight:
238.41
Beilstein:
8701742
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

£122.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCC1CCC1=O

InChI

1S/C16H30O/c1-2-3-4-5-6-7-8-9-10-11-12-15-13-14-16(15)17/h15H,2-14H2,1H3

InChI key

VVKRFZDUZMLMQM-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mostafa Taghvaei et al.
Journal of food science, 85(8), 2608-2614 (2020-07-08)
The food irradiation marker, 2-dodecylcyclobutanone (2-DCB), assayed by SPME provides a fast and simple method to estimate the irradiation history of fat-containing food products. The SPME conditions were optimized to maximize the extraction of 2-DCB from chicken jerky treats (CJT)
Andreas Breidbach et al.
Food chemistry, 201, 52-58 (2016-02-13)
Irradiation of food products and ingredients must be indicated by proper labeling. This study evaluated the appropriateness of the European Standard EN 1785:2003 for the detection of 2-alkylcyclobutanones, which are radiolysis products of fatty acids, in cashew nuts and nutmeg
Ki-Chang Lee et al.
Journal of agricultural and food chemistry, 68(9), 2803-2815 (2020-02-11)
This study evaluated the applicability of a rapid analytical method using a headspace solid-phase microextraction gas chromatography/mass spectrometry (HS-SPME-GC/MS) technique to identify gamma-irradiated soybeans (0.1-5 kGy). From the partial least squares discriminant analysis used to discriminate between non-irradiated and irradiated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service