Skip to Content
Merck
All Photos(1)

Key Documents

40647

Supelco

Allyl sulfide

analytical standard

Synonym(s):

3,3′-Thiobis-1-propene, Diallyl sulfide, NSC 20947

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2=CHCH2)2S
CAS Number:
Molecular Weight:
114.21
Beilstein:
1736016
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

vapor density

3.9 (vs air)

vapor pressure

7 mmHg ( 20 °C)

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.490 (lit.)

bp

138 °C (lit.)

mp

−83 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

C=CCSCC=C

InChI

1S/C6H10S/c1-3-5-7-6-4-2/h3-4H,1-2,5-6H2

InChI key

UBJVUCKUDDKUJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiangsheng Xiao et al.
PloS one, 9(11), e112720-e112720 (2014-11-15)
Diallyl disulfide (DADS) is one of the major volatile components of garlic oil. DADS has various biological properties, including anticancer, antiangiogenic, and antioxidant effects. However, the anticancer mechanisms of DADS in human breast cancer have not been elucidated, particularly in
Bo Su et al.
Oncology reports, 33(5), 2484-2494 (2015-03-31)
Diallyl disulfide (DADS) is characterized as an effective agent for the prevention and therapy of cancer, however, mechanisms regarding its anticancer effects are not fully clarified. In the present study, we compared the protein expression profile of gastric cancer MGC-803
Israr Ahmad et al.
Molecular and cellular biochemistry, 393(1-2), 209-222 (2014-04-29)
Cytochrome P4502E1 (CYP2E1), glutathione-S-transferase A4-4 (GSTA4-4), and inducible nitric oxide synthase (iNOS) are implicated in maneb- and paraquat-induced toxicity leading to various pathological conditions. The study aimed to investigate the role of CYP2E1 in maneb- and paraquat-induced oxidative stress in
Joan Eilstein et al.
Archives of toxicology, 88(9), 1681-1694 (2014-03-25)
Skin function is not limited to a physical barrier. According to its total surface area, it is also considered as an extra-hepatic metabolizing organ. In vitro engineered human skins have been developed to replace limited ex vivo normal human skin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service