Skip to Content
Merck
All Photos(1)

Documents

38863

Supelco

2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-methyl-L-valinate hydrochloride

pharmaceutical impurity standard

Synonym(s):

Valaciclovir Related Compound C (USP), Valaciclovir impurity C (PhEur), N-Methyl-L-valine 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N6O4 · HCl
CAS Number:
Molecular Weight:
374.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10% related substances HPLC impurities
≤5.0% residual solvents
≤5.0 wt. % water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN[C@@H](C(C)C)C(=O)OCCOCn1cnc2C(=O)N=C(N)Nc12

InChI

1S/C14H22N6O4.ClH/c1-8(2)9(16-3)13(22)24-5-4-23-7-20-6-17-10-11(20)18-14(15)19-12(10)21;/h6,8-9,16H,4-5,7H2,1-3H3,(H3,15,18,19,21);1H/t9-;/m0./s1

InChI key

RLJNMIUABOJDNS-FVGYRXGTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service