Skip to Content
Merck
All Photos(1)

Documents

33943

Supelco

Dimepiperate

PESTANAL®, analytical standard

Synonym(s):

S-(1-Methyl-1-phenylethyl)-1-piperidinecarbothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NOS
CAS Number:
Molecular Weight:
263.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(SC(=O)N1CCCCC1)c2ccccc2

InChI

1S/C15H21NOS/c1-15(2,13-9-5-3-6-10-13)18-14(17)16-11-7-4-8-12-16/h3,5-6,9-10H,4,7-8,11-12H2,1-2H3

InChI key

BWUPSGJXXPATLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Dimepiperate is a systemic, thiocarbamate herbicide used for the control of barnyard grass in flooded rice fields.

Application

Dimepiperate may be used as an analytical reference standard for the determination of the analyte in:
  • Shellfish using gas chromatography coupled to tandem mass spectrometry (GC/MS/MS).
  • Rice paddy fields using quick easy cheap effective rugged safe (QuEChERS) method combined with gas chromatography coupled to mass spectrometry (GC-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia, 619(1), 67-74 (2014)
A multi-residue method for the determination of 203 pesticides in rice paddies using gas chromatography/mass spectrometry.
Nguyen TD, et al.
Analytica Chimica Acta, 619(1), 67-74 (2008)
Analysis of banned veterinary drugs and herbicide residues in shellfish by liquid chromatography-tandem mass spectrometry (LC/MS/MS) and gas chromatography-tandem mass spectrometry (GC/MS/MS).
Chang GR, et al.
Marine Pollution Bulletin, 113(1-2), 579-584 (2016)
E Hakme et al.
Chemosphere, 184, 1310-1319 (2017-07-07)
This work presents a non-targeted screening approach for the detection and quantitation of contaminants in bees and pollen, collected from the same hive, by GC-EI-ToF-MS. It consists of a spectral library datasets search using a compound database followed by a
Jishi Wang et al.
Journal of chromatography. A, 1521, 10-18 (2017-09-25)
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service