Skip to Content
Merck
All Photos(1)

Key Documents

33830

Sigma-Aldrich

Dibenzyl sulfoxide

purum, ≥98.0% (HPLC)

Synonym(s):

Benzyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2SO
CAS Number:
Molecular Weight:
230.33
Beilstein:
2049262
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (HPLC)

mp

130-132 °C (lit.)
130-132 °C

SMILES string

O=S(Cc1ccccc1)Cc2ccccc2

Gene Information

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service