Skip to Content
Merck
All Photos(1)

Documents

32564

Supelco

Butyl 4-hydroxybenzoate-ring-13C6 solution

50 μg/mL in acetone, analytical standard

Synonym(s):

4-Hydroxybenzoic acid n-butyl ester-ring-13C6 solution, Butyl paraben-ring-13C6 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C5H14O3
CAS Number:
Molecular Weight:
200.18
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+5

storage temp.

−20°C

SMILES string

CCCCOC(=O)[13c]1[13cH][13cH][13c](O)c([13cH]1)C#CC#CC#CC#CC#CC#C

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service