Skip to Content
Merck
All Photos(1)

Key Documents

29875

Sigma-Aldrich

Cyclophosphamide monohydrate

Synonym(s):

2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, Cytoxan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15Cl2N2O2P · H2O
CAS Number:
Molecular Weight:
279.10
Beilstein:
4678992
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

mp

49-51 °C (lit.)

solubility

H2O: 0.1 g/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[H]O[H].ClCCN(CCCl)P1(=O)NCCCO1

InChI

1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2

InChI key

PWOQRKCAHTVFLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cyclophosphamide is a cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations. Its clinical activity is associated with a decrease in aldehyde dehydrogenase 1 (ALDH1) activity.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Muta. 1B - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Pugnet et al.
Clinical and experimental rheumatology, 31(1 Suppl 75), S62-S64 (2013-07-24)
Progressive multifocal encephalopathy (PML) is a rare demyelinating disorder targeting the central nervous system and resulting from JC virus reactivation. PML occurs in patients immunocompromised because of haematological malignancies, HIV infection or treatment with cytotoxic drugs. Herein, we describe PML
Jennifer L Guerriero et al.
Cancer research, 68(23), 9595-9600 (2008-12-03)
Dysregulation of apoptosis is associated with the development of human cancer and resistance to anticancer therapy. The ultimate goal of cancer treatment is to selectively induce cancer cell death and overcome drug resistance. A deeper understanding of how a given
Parotitis as the presenting symptom of Wegener's granulomatosis: case report and meta-analysis.
Ilan Green et al.
The Israel Medical Association journal : IMAJ, 15(3), 188-192 (2013-05-15)
Ulrich Specks et al.
The New England journal of medicine, 369(5), 417-427 (2013-08-02)
The 18-month efficacy of a single course of rituximab as compared with conventional immunosuppression with cyclophosphamide followed by azathioprine in patients with severe (organ-threatening) antineutrophil cytoplasmic antibody (ANCA)-associated vasculitis is unknown. In a multicenter, randomized, double-blind, double-dummy, noninferiority trial, we
Z-H Niu et al.
European review for medical and pharmacological sciences, 17(10), 1337-1344 (2013-06-07)
Preoperative chemotherapy is often used in patients with locally advanced breast cancer. However, commonly used clinical and pathological parameters are poor predictors of response to this type of therapy. The secreted form of the CLU protein (sCLU) is a glycosylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service